missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
Solubility
Soluble in DMSO.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
Solubility
Soluble in DMSO.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | >300°C |
Odor | Odorless |
Quantity | 250 mg |
Beilstein | 139956 |
Solubility Information | Soluble in DMSO. |
Formula Weight | 152.11 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'