missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, 98%, ACROS Organics™

Brand:  Acros Organics 163030250

Code : 29

Additional Details : CAS Number : 6299-25-8 Weight : 0.07500kg Transport : UN number : 3263 Chem class : 1b Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10791355

  • £150.70 / 25g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

4, 6-Dichloro-2-(methylthio)pyrimidine
97.5
98%
C5H4Cl2N2S
MFCD00006086
6299-25-8
100.0
98%
195.07

Specifications

4, 6-Dichloro-2-(methylthio)pyrimidine
135.0° to 136.0°C (14.0 mmHg)
38.0° to 42.0°C
6299-25-8
100.0
Powder or Crystals
98%
195.07
Glass bottle
>112°C
White to yellow or orange
25g
97.5
Authentic
98%
C5H4Cl2N2S
MFCD00006086
23, 371
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4,6-Dichloro-2-(methylthio)pyrimidine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS