missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-3736195
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6299-25-8
C5H4Cl2N2S
195.07
MFCD00006086
FCMLONIWOAGZJX-UHFFFAOYSA-N
4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
80531
4,6-dichloro-2-methylsulfanylpyrimidine
CSC1=NC(=CC(=N1)Cl)Cl
This item is not returnable. View return policy
6299-25-8
C5H4Cl2N2S
195.07
MFCD00006086
FCMLONIWOAGZJX-UHFFFAOYSA-N
4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
80531
4,6-dichloro-2-methylsulfanylpyrimidine
CSC1=NC(=CC(=N1)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C to 42.0°C
Color Orange or White to Yellow
Boiling Point 135.0°C to 136.0°C (14.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 371
Formula Weight 195.07
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4, 6-Dichloro-2-(methylthio)pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dichloro-2-(methylthio)pyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'