missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminoresorcinol dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 10663572
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16523-31-2
C6H8N2O2·2HCl
213.06
MFCD00143239
KUMOYHHELWKOCB-UHFFFAOYSA-N
4,6-diaminoresorcinol dihydrochloride, 4,6-diaminobenzene-1,3-diol dihydrochloride, 4,6-diaminoresorcinol 2hcl, 1,3-benzenediol, 4,6-diamino-, dihydrochloride, 4,6-diamino resorcinol dihydrochloride, 4,6-diamino-benzene-1,3-diol 2hcl salt, 1,3-benzenediol, 4,6-diamino-, hydrochloride 1:2, 4,6-bis azanyl benzene-1,3-diol dihydrochloride, pubchem20873, acmc-1c78d
2733648
4,6-diaminobenzene-1,3-diol;dihydrochloride
C1=C(C(=CC(=C1N)O)O)N.Cl.Cl
This item is not returnable. View return policy
16523-31-2
C6H8N2O2·2HCl
213.06
MFCD00143239
KUMOYHHELWKOCB-UHFFFAOYSA-N
4,6-diaminoresorcinol dihydrochloride, 4,6-diaminobenzene-1,3-diol dihydrochloride, 4,6-diaminoresorcinol 2hcl, 1,3-benzenediol, 4,6-diamino-, dihydrochloride, 4,6-diamino resorcinol dihydrochloride, 4,6-diamino-benzene-1,3-diol 2hcl salt, 1,3-benzenediol, 4,6-diamino-, hydrochloride 1:2, 4,6-bis azanyl benzene-1,3-diol dihydrochloride, pubchem20873, acmc-1c78d
2733648
4,6-diaminobenzene-1,3-diol;dihydrochloride
C1=C(C(=CC(=C1N)O)O)N.Cl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 13,I,317
Formula Weight 213.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4, 6-Diaminoresorcinol dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Diaminoresorcinol dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'