missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminopyrimidine Hemisulfate, Monohydrate 98%, ACROS Organics™

Brand:  Acros Organics 112410010

Code : 29

Additional Details : CAS Number : 77709-02-5 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10615342

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4, 6-Diaminopyrimidine hemisulfate monohydrate
77709-02-5
100.0
98%
177.18
98%
97.5
98%
5 H2SO4·H2O
MFCD00149655

Specifications

4, 6-Diaminopyrimidine hemisulfate monohydrate
243.0° to 244.0°C
98%
97.5
Authentic
Crystalline Powder
98%
177.18
Yellow
1g
77709-02-5
100.0
Glass bottle
98%
5 H2SO4·H2O
MFCD00149655
Videos
SDS