Learn More
Thermo Scientific Maybridge 4-(6-Bromopyrid-2-yl)morpholine, 1g, 97%
Brand: Thermo Scientific Maybridge CC38310DA
4-(6-Bromopyrid-2-yl)morpholine, 1g, 97%
Code : 31
Additional Details : CAS Number : 332134-60-8 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III