missing translation for 'onlineSavingsMsg'
Learn More

4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7029820
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
261733-18-0
C36H27NOP2
551.57
MFCD03788937
HSWZLYXRAOXOLL-UHFFFAOYSA-N
4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
3255346
(6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane
N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
261733-18-0
C36H27NOP2
551.57
MFCD03788937
HSWZLYXRAOXOLL-UHFFFAOYSA-N
4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
3255346
(6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane
N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 251°C to 254°C
Quantity 250 mg
Solubility Information Miscible with water.
Formula Weight 551.55
Percent Purity ≥98%
Chemical Name or Material 4,6-Bis(diphenylphosphino)phenoxazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'