missing translation for 'onlineSavingsMsg'
Learn More

4-[5-(Trifluoromethyl)pyridin-2-yl]benzaldehyde, 90%, Thermo Scientific™

Catalog Number p-3522525
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
871252-64-1
C13H8F3NO
251.208
MFCD06797372
SBXWMOPSHGIWEN-UHFFFAOYSA-N
4-5-trifluoromethyl pyridin-2-yl benzaldehyde, 4-5-trifluoromethyl-2-pyridinyl benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridinyl-benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridyl benzaldehyde, 4-5-trifluoromethyl-pyridin-2-yl-benzaldehyde, benzaldehyde,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl pyridin-2-yl benzenecarbaldehyde
2763359
4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
C1=CC(=CC=C1C=O)C2=NC=C(C=C2)C(F)(F)F
This item is not returnable. View return policy
871252-64-1
C13H8F3NO
251.208
MFCD06797372
SBXWMOPSHGIWEN-UHFFFAOYSA-N
4-5-trifluoromethyl pyridin-2-yl benzaldehyde, 4-5-trifluoromethyl-2-pyridinyl benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridinyl-benzenecarbaldehyde, 4-5-trifluoromethyl-2-pyridyl benzaldehyde, 4-5-trifluoromethyl-pyridin-2-yl-benzaldehyde, benzaldehyde,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl pyridin-2-yl benzenecarbaldehyde
2763359
4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
C1=CC(=CC=C1C=O)C2=NC=C(C=C2)C(F)(F)F
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 251.21
Percent Purity 90%
Chemical Name or Material 4-[5-(Trifluoromethyl)pyridin-2-yl]benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'