missing translation for 'onlineSavingsMsg'
Learn More

{4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methanol, 97%, Thermo Scientific™

Catalog Number p-3541189
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
613239-75-1
C13H10F3NO
253.22
MFCD09817477
VRVOASJNEFKGGI-UHFFFAOYSA-N
4-5-trifluoromethyl pyridin-2-yl phenyl methanol, 4-5-trifluoromethyl pyridin-2-yl benzyl alcohol, 4-5-trifluoromethyl-2-pyridinyl phenyl methanol, 4-5-trifluoromethyl pyrid-2-yl phenyl methanol, 4-5-trifluoromethyl-pyridin-2-yl-phenyl-methanol, 4-5-trifluoromethyl-2-pyridyl phenyl methan-1-ol
22280022
[4-[5-(trifluoromethyl)pyridin-2-yl]phenyl]methanol
OCC1=CC=C(C=C1)C1=NC=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
613239-75-1
C13H10F3NO
253.22
MFCD09817477
VRVOASJNEFKGGI-UHFFFAOYSA-N
4-5-trifluoromethyl pyridin-2-yl phenyl methanol, 4-5-trifluoromethyl pyridin-2-yl benzyl alcohol, 4-5-trifluoromethyl-2-pyridinyl phenyl methanol, 4-5-trifluoromethyl pyrid-2-yl phenyl methanol, 4-5-trifluoromethyl-pyridin-2-yl-phenyl-methanol, 4-5-trifluoromethyl-2-pyridyl phenyl methan-1-ol
22280022
[4-[5-(trifluoromethyl)pyridin-2-yl]phenyl]methanol
OCC1=CC=C(C=C1)C1=NC=C(C=C1)C(F)(F)F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 253.22
Percent Purity 97%
Chemical Name or Material {4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'