missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3629700
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
179056-82-7
C10H8N2O2
188.19
MFCD11109323
MPECIJQJPPLBGV-UHFFFAOYSA-N
4-5-methyl-1,3,4-oxadiazol-2-yl benzaldehyde, 2-4-formylphenyl-5-methyl-1,3,4-oxadiazole, benzaldehyde,4-5-methyl-1,3,4-oxadiazol-2-yl, 4-5-methyl-1,3,4-oxadiazol-2-yl benzaldehyd, benzaldehyde, 4-5-methyl-1,3,4-oxadiazol-2-yl
10655207
4-(5-methyl-1,3,4-oxadiazol-2-yl)benzaldehyde
CC1=NN=C(O1)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
179056-82-7
C10H8N2O2
188.19
MFCD11109323
MPECIJQJPPLBGV-UHFFFAOYSA-N
4-5-methyl-1,3,4-oxadiazol-2-yl benzaldehyde, 2-4-formylphenyl-5-methyl-1,3,4-oxadiazole, benzaldehyde,4-5-methyl-1,3,4-oxadiazol-2-yl, 4-5-methyl-1,3,4-oxadiazol-2-yl benzaldehyd, benzaldehyde, 4-5-methyl-1,3,4-oxadiazol-2-yl
10655207
4-(5-methyl-1,3,4-oxadiazol-2-yl)benzaldehyde
CC1=NN=C(O1)C1=CC=C(C=O)C=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 4-(5-Methyl-1, 3, 4-oxadiazol-2-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'