missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Imidazoledicarboxylic acid, 99%, ACROS Organics™
£92.22
Chemical Identifiers
CAS | 570-22-9 |
---|---|
Molecular Formula | C5H4N2O4 |
Molecular Weight (g/mol) | 156.1 |
MDL Number | MFCD00005200 |
InChI Key | ZEVWQFWTGHFIDH-UHFFFAOYSA-N |
Synonym | 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid |
PubChem CID | 68442 |
IUPAC Name | 1H-imidazole-4,5-dicarboxylic acid |
SMILES | C1=NC(=C(N1)C(=O)O)C(=O)O |
Chemical Identifiers
570-22-9 | |
156.1 | |
ZEVWQFWTGHFIDH-UHFFFAOYSA-N | |
68442 | |
C1=NC(=C(N1)C(=O)O)C(=O)O |
C5H4N2O4 | |
MFCD00005200 | |
4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid | |
1H-imidazole-4,5-dicarboxylic acid |
Specifications
4, 5-Imidazoledicarboxylic acid | |
Glass bottle | |
265.0°C | |
99% | |
98.5 | |
99% | |
MFCD00005200 | |
Solubility in water: 0.5g/L (20°C) | |
C1=NC(=C(N1)C(=O)O)C(=O)O | |
156.1 | |
156.1 | |
99% |
Authentic | |
Brown-Yellow to Yellow | |
25g | |
570-22-9 | |
100.0 | |
C5H4N2O4 | |
4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid | |
ZEVWQFWTGHFIDH-UHFFFAOYSA-N | |
1H-imidazole-4,5-dicarboxylic acid | |
68442 | |
Fine Crystalline Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4,5-Imidazoledicarboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.