missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dimethyl-2-furaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.0100g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 4,5-dimethyl-2-furaldehyde, 4,5-dimethylfurfural, 4,5-dimethyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 4,5-dimethyl, 2-furanmethanethiolformate, acmc-1ao31, 4.5-dimethyl-2-furaldehyde, 4,5-dimethyl-2-furancarbaldehyde, 4,5-dimethyl-2-furancarboxaldehyde, 2-furancarboxaldehyde,4,5-dimethyl |
Formula Weight | 124.14 |
Percent Purity | 97% |
Chemical Name or Material | 4, 5-Dimethyl-2-furaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'