missing translation for 'onlineSavingsMsg'
Learn More

Croconic acid, 97%, Acros Organics™

Brand:  Acros Organics 374935000

Code : 29

Additional Details : CAS Number : 488-86-8 Weight : 0.05050kg


Quantity 500mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10220543

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4,5-Dihydroxy-4-cyclopentene-1,2,3-trione
96.0
≥96.0%
C5H2O5
MFCD00181389
488-86-8
100.0
97%
142.07
Croconic acid

Specifications

4,5-Dihydroxy-4-cyclopentene-1,2,3-trione
500mg
96.0
Authentic
Powder or Crystals
97%
142.07
Croconic acid
Yellow
488-86-8
100.0
Glass bottle
≥96.0%
C5H2O5
MFCD00181389
Videos
SDS