missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-methoxyphenylacetic acid, 97%, Thermo Scientific™

Catalog Number p-7029210
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886761-73-5
C9H8F2O3
202.157
MFCD06660348
UYEGFRSSHGRVJF-UHFFFAOYSA-N
4,5-difluoro-2-methoxyphenylacetic acid, 2-4,5-difluoro-2-methoxyphenyl acetic acid, 2-3,4-difluoro-6-methoxyphenyl acetic acid, 4,5-difluoro-2-methoxyphenyl acetic acid
17750767
2-(4,5-difluoro-2-methoxyphenyl)acetic acid
COC1=CC(=C(C=C1CC(=O)O)F)F
This item is not returnable. View return policy
886761-73-5
C9H8F2O3
202.157
MFCD06660348
UYEGFRSSHGRVJF-UHFFFAOYSA-N
4,5-difluoro-2-methoxyphenylacetic acid, 2-4,5-difluoro-2-methoxyphenyl acetic acid, 2-3,4-difluoro-6-methoxyphenyl acetic acid, 4,5-difluoro-2-methoxyphenyl acetic acid
17750767
2-(4,5-difluoro-2-methoxyphenyl)acetic acid
COC1=CC(=C(C=C1CC(=O)O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 202.16
Percent Purity 97%
Chemical Name or Material 4,5-Difluoro-2-methoxyphenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'