missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,5-Difluoro-2-iodobenzoic acid, 98%

Brand:  Alfa Aesar™ H61798.03

UNSPSC : 12352106

Code : NEW

Additional Details : CAS Number : 130137-05-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418378

  • £119.78 / 1g


Estimated Shipment date
from Supplier 13-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

4,5-Difluoro-2-iodobenzoic acid
130137-05-2
284
MFCD02684316

Specifications

4,5-Difluoro-2-iodobenzoic acid
1g
Solid
284
Light Sensitive
130137-05-2
98%
MFCD02684316
Documentation

Technical Documents