missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,5-Difluoro-2-iodobenzoic acid, 98%

Brand:  Alfa Aesar™ H61798.06

Code : NEW

Additional Details : CAS Number : 130137-05-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428378

  • £597.84 / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

4,5-Difluoro-2-iodobenzoic acid
130137-05-2
284
MFCD02684316

Specifications

4,5-Difluoro-2-iodobenzoic acid
5g
Solid
284
Light Sensitive
130137-05-2
98%
MFCD02684316
Videos
SDS