missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichlorophthalic anhydride, 98%, Thermo Scientific Chemicals

Catalog Number 10686824
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
942-06-3
C8H2Cl2O3
217.00
MFCD00075034
ULSOWUBMELTORB-UHFFFAOYSA-N
4,5-dichlorophthalic anhydride, 5,6-dichloroisobenzofuran-1,3-dione, 4,5-dichlorophthalicanhydride, 1,3-isobenzofurandione, 5,6-dichloro, 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione, 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 5,6-dichlor-2-benzofuran-1,3-dion, pubchem16761, acmc-209rqk, 4,5-dichlorophalic anhydride
70334
5,6-dichloro-2-benzofuran-1,3-dione
ClC1=CC2=C(C=C1Cl)C(=O)OC2=O
This item is not returnable. View return policy
942-06-3
C8H2Cl2O3
217.00
MFCD00075034
ULSOWUBMELTORB-UHFFFAOYSA-N
4,5-dichlorophthalic anhydride, 5,6-dichloroisobenzofuran-1,3-dione, 4,5-dichlorophthalicanhydride, 1,3-isobenzofurandione, 5,6-dichloro, 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione, 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 5,6-dichlor-2-benzofuran-1,3-dion, pubchem16761, acmc-209rqk, 4,5-dichlorophalic anhydride
70334
5,6-dichloro-2-benzofuran-1,3-dione
ClC1=CC2=C(C=C1Cl)C(=O)OC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 185°C to 187°C
Color Blue to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 217.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 5-Dichlorophthalic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dichlorophthalic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'