missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloroisothiazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10713233
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
131947-13-2
C4HCl2NO2S
198.017
MFCD00186468
ZFEHQZVNKOESSZ-UHFFFAOYSA-N
1244565
4,5-dichloro-1,2-thiazole-3-carboxylic acid
C1(=C(SN=C1C(=O)O)Cl)Cl
This item is not returnable. View return policy
131947-13-2
C4HCl2NO2S
198.017
MFCD00186468
ZFEHQZVNKOESSZ-UHFFFAOYSA-N
1244565
4,5-dichloro-1,2-thiazole-3-carboxylic acid
C1(=C(SN=C1C(=O)O)Cl)Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 4,5-dichloroisothiazole-3-carboxylic acid, 4,5-dichloro-isothiazole-3-carboxylic acid, 3-isothiazolecarboxylicacid, 4,5-dichloro, dichloro-1,2-thiazole-3-carboxylic acid, acmc-1bwzv, 3-isothiazolecarboxylicacid,4,5-dichloro, 4,5-dichloro-3-isothiazolecarboxylic acid
Formula Weight 198.03
Percent Purity 97%
Chemical Name or Material 4, 5-Dichloroisothiazole-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'