missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dichloroisothiazole-3-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Synonym | 4,5-dichloroisothiazole-3-carboxylic acid, 4,5-dichloro-isothiazole-3-carboxylic acid, 3-isothiazolecarboxylicacid, 4,5-dichloro, dichloro-1,2-thiazole-3-carboxylic acid, acmc-1bwzv, 3-isothiazolecarboxylicacid,4,5-dichloro, 4,5-dichloro-3-isothiazolecarboxylic acid |
Formula Weight | 198.03 |
Percent Purity | 97% |
Chemical Name or Material | 4, 5-Dichloroisothiazole-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'