missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Diazafluoren-9-one, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.
Specifications
Melting Point | 214°C to 217°C |
Quantity | 5 g |
Solubility Information | Soluble in dichloromethane,tetrahydrofuran,chloroform,benzene,toluene and polar organic solvents. Insoluble in diethyl ether and alkanes. |
Formula Weight | 182.18 |
Percent Purity | 98% |
Chemical Name or Material | 4,5-Diazafluoren-9-one |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'