missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridyl)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7036964
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
99163-12-9
C12H9NO
183.21
MFCD02684110
MBJXDIYHLGBQOT-UHFFFAOYSA-N
4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt
639673
4-pyridin-4-ylbenzaldehyde
C1=CC(=CC=C1C=O)C2=CC=NC=C2
This item is not returnable. View return policy
99163-12-9
C12H9NO
183.21
MFCD02684110
MBJXDIYHLGBQOT-UHFFFAOYSA-N
4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt
639673
4-pyridin-4-ylbenzaldehyde
C1=CC(=CC=C1C=O)C2=CC=NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 85°C to 89°C
Quantity 250 mg
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-(4-Pyridyl)benzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'