missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridyl)-2-(2-pyridylmethyl)thiazole, 97%, Thermo Scientific™

Catalog Number p-7039580
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1421263-46-8
C14H11N3S
253.323
MFCD22683021
RFXGELSJVHDCRI-UHFFFAOYSA-N
4-2-pyridin-2-ylmethyl-1,3-thiazol-4-yl pyridine, 4-4-pyridyl-2-2-pyridylmethyl thiazole
97030365
4-pyridin-4-yl-2-(pyridin-2-ylmethyl)-1,3-thiazole
C1=CC=NC(=C1)CC2=NC(=CS2)C3=CC=NC=C3
This item is not returnable. View return policy
1421263-46-8
C14H11N3S
253.323
MFCD22683021
RFXGELSJVHDCRI-UHFFFAOYSA-N
4-2-pyridin-2-ylmethyl-1,3-thiazol-4-yl pyridine, 4-4-pyridyl-2-2-pyridylmethyl thiazole
97030365
4-pyridin-4-yl-2-(pyridin-2-ylmethyl)-1,3-thiazole
C1=CC=NC(=C1)CC2=NC(=CS2)C3=CC=NC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 253.32
Percent Purity 97%
Chemical Name or Material 4-(4-Pyridyl)-2-(2-pyridylmethyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'