missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,4'-Oxybis(benzenesulfonyl hydrazide), 95%

£63.59 - £230.66

Chemical Identifiers

CAS 80-51-3
Molecular Formula C12H14N4O5S2
Molecular Weight (g/mol) 358.387
MDL Number MFCD00007587
InChI Key NBOCQTNZUPTTEI-UHFFFAOYSA-N
Synonym 4,4'-oxydibenzenesulfonohydrazide, 4,4'-oxybis benzenesulfonyl hydrazide, celogen, serogen, obsh, genitron ob, zhenitron ov, celogen ot, 4,4'-oxybis benzenesulfonhydrazide, cellmic s
PubChem CID 6649
IUPAC Name 4-[4-(hydrazinesulfonyl)phenoxy]benzenesulfonohydrazide
SMILES C1=CC(=CC=C1OC2=CC=C(C=C2)S(=O)(=O)NN)S(=O)(=O)NN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427748
SDS View Documents
Alfa Aesar™
H61239.22
100g £63.59
100g View alternative products
Save Up to 
»
Estimated Shipment
23-10-2020
Please to purchase this item. Need a web account? Register with us today!
15437748
SDS View Documents
Alfa Aesar™
H61239.36
500g £230.66
500g View alternative products
Save Up to 
»
Estimated Shipment
23-10-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
4,4′-Oxybis(benzenesulfonyl hydrazide) is an organic hydrazide used as a blowing agent for sponge rubber and expanded plastics. Used as an additive to polymers to make sponge rubber cosmetic puffs, electrostatographic toners, buoyant seats, insulation and packing materials, adhesives, sealants, and leather substitutes.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

80-51-3
358.387
NBOCQTNZUPTTEI-UHFFFAOYSA-N
6649
C1=CC(=CC=C1OC2=CC=C(C=C2)S(=O)(=O)NN)S(=O)(=O)NN
C12H14N4O5S2
MFCD00007587
4,4'-oxydibenzenesulfonohydrazide, 4,4'-oxybis benzenesulfonyl hydrazide, celogen, serogen, obsh, genitron ob, zhenitron ov, celogen ot, 4,4'-oxybis benzenesulfonhydrazide, cellmic s
4-[4-(hydrazinesulfonyl)phenoxy]benzenesulfonohydrazide

Specifications

4,4'-Oxybis(benzenesulfonyl hydrazide)
100g
C12H14N4O5S2
UN3226
NBOCQTNZUPTTEI-UHFFFAOYSA-N
4-[4-(hydrazinesulfonyl)phenoxy]benzenesulfonohydrazide
6649
Solid
White
80-51-3
MFCD00007587
4,4'-oxydibenzenesulfonohydrazide, 4,4'-oxybis benzenesulfonyl hydrazide, celogen, serogen, obsh, genitron ob, zhenitron ov, celogen ot, 4,4'-oxybis benzenesulfonhydrazide, cellmic s
C1=CC(=CC=C1OC2=CC=C(C=C2)S(=O)(=O)NN)S(=O)(=O)NN
358.387
358.39
95%