missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Nitrobenzyl)pyridine, 98+%, Thermo Scientific Chemicals
18.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids.
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Melting Point | 69°C to 72°C |
Quantity | 5 g |
Beilstein | 170877 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in acetone. Insoluble in water. |
Formula Weight | 214.22 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-(4-Nitrobenzyl)pyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'