missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, 97%

£43.90 - £150.00

Chemical Identifiers

CAS 656239-38-2
Molecular Formula C17H24BNO4
Molecular Weight (g/mol) 317.19
MDL Number MFCD05863913
InChI Key ZOPBEWUNCNLYSD-UHFFFAOYSA-N
Synonym morpholino 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-morpholinocarbonyl phenylboronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 4-morpholine carboxamidophenylboronic acid, pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 4-morpholine-4-carbonyl phenyl boronic acid pinacol ester
PubChem CID 2760141
IUPAC Name morpholin-4-yl-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11432060
SDS View Documents
Alfa Aesar™
H28484.03
1g
£43.90
1g
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2021
Please to purchase this item. Need a web account? Register with us today!
11442060
SDS View Documents
Alfa Aesar™
H28484.06
5g
£150.00
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

656239-38-2
317.19
ZOPBEWUNCNLYSD-UHFFFAOYSA-N
2760141
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1
C17H24BNO4
MFCD05863913
morpholino 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-morpholinocarbonyl phenylboronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 4-morpholine carboxamidophenylboronic acid, pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 4-morpholine-4-carbonyl phenyl boronic acid pinacol ester
morpholin-4-yl-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone

Specifications

4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
1g
C17H24BNO4
morpholino 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-morpholinocarbonyl phenylboronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 4-morpholine carboxamidophenylboronic acid, pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 4-morpholine-4-carbonyl phenyl boronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1
317.19
317.19
187°C to 193°C
656239-38-2
MFCD05863913
ZOPBEWUNCNLYSD-UHFFFAOYSA-N
morpholin-4-yl-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
2760141
97%
SDS
Product Certifications