missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11432060
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
656239-38-2
C17H24BNO4
317.19
MFCD05863913
ZOPBEWUNCNLYSD-UHFFFAOYSA-N
morpholino 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-morpholinocarbonyl phenylboronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 4-morpholine carboxamidophenylboronic acid, pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 4-morpholine-4-carbonyl phenyl boronic acid pinacol ester
2760141
morpholin-4-yl-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1
This item is not returnable. View return policy
656239-38-2
C17H24BNO4
317.19
MFCD05863913
ZOPBEWUNCNLYSD-UHFFFAOYSA-N
morpholino 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-morpholinocarbonyl phenylboronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 4-morpholine carboxamidophenylboronic acid, pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 4-morpholine-4-carbonyl phenyl boronic acid pinacol ester
2760141
morpholin-4-yl-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 187°C to 193°C
Quantity 1 g
Formula Weight 317.19
Percent Purity 97%
Chemical Name or Material 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'