missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Morpholinyl)aniline, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
Description
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
Solubility
Soluble in chloroform, ethyl acetate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
Solubility
Soluble in chloroform, ethyl acetate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
Specifications
Melting Point | 129°C to 133°C |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 150957 |
Solubility Information | Soluble in chloroform,ethyl acetate. |
Formula Weight | 178.24 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-(4-Morpholinyl)aniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'