4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC45616DA

1GR 4-¢(4-Methylpiperazin-1-yl)methyl!benzonitrile, 97%

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 125743-63-7 Weight: 0.05100kg Transport: UN number : 2922 Chem class : 1b Pack group : III


Packaging Amber glass bottle
Quantity 1g

Product Code. 10760544

Quantity Price
1 £ 29.0 / 1g
Estimated Shipment date
from Supplier 21-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile
CAS 125743-63-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17N3
Formula Weight 215.3
MDL Number MFCD08060531

Specification

Formula Weight 215.3
Quantity 1g
Assay 97%
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 125743-63-7
Assay Percent Range 97%
MDL Number MFCD08060531
Packaging Amber glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Molecular Formula C13H17N3
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzonitrile
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.