missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochlorideHemihydrate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 316.23 |
Percent Purity | 95% |
Chemical Name or Material | 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride 0.5 hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'