missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenylethynyl)benzeneboronic acid pinacol ester,95%, Thermo Scientific™

Catalog Number 15469405
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
1190376-24-9
C21H23BO3
334.22
MFCD16294533
RRYKLXOOBFDYDY-UHFFFAOYSA-N
2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, 2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb224, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
71306524
COC1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
1190376-24-9
C21H23BO3
334.22
MFCD16294533
RRYKLXOOBFDYDY-UHFFFAOYSA-N
2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, 2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb224, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
71306524
COC1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
IUPAC Name 2-{4-[2-(4-methoxyphenyl)ethynyl]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula Weight 334.22
Percent Purity 95%
Chemical Name or Material 4-(4-Methoxyphenylethynyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'