missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.
Solubility
Slightly soluble in methanol. Insoluble in chloroform.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.
Solubility
Slightly soluble in methanol. Insoluble in chloroform.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 56°C to 59°C |
Quantity | 1 g |
Beilstein | 2416568 |
Solubility Information | Slightly soluble in methanol. Insoluble in chloroform. |
Formula Weight | 194.23 |
Percent Purity | 98% |
Chemical Name or Material | 4-(4-Methoxyphenyl)butyric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'