missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)-2-butanone, 99%, Thermo Scientific Chemicals

Catalog Number p-7081162
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
104-20-1
C11H14O2
178.23
MFCD00008791
PCBSXBYCASFXTM-UHFFFAOYSA-N
4-4-methoxyphenyl-2-butanone, anisylacetone, 4-4-methoxyphenyl butan-2-one, 4-methoxybenzylacetone, raspberry ketone methyl ether, 2-butanone, 4-4-methoxyphenyl, anisylacetone, p, 4-p-methoxyphenyl-2-butanone, methyl oxanone, 1-4-methoxyphenyl-3-butanone
61007
4-(4-methoxyphenyl)butan-2-one
COC1=CC=C(CCC(C)=O)C=C1
This item is not returnable. View return policy
104-20-1
C11H14O2
178.23
MFCD00008791
PCBSXBYCASFXTM-UHFFFAOYSA-N
4-4-methoxyphenyl-2-butanone, anisylacetone, 4-4-methoxyphenyl butan-2-one, 4-methoxybenzylacetone, raspberry ketone methyl ether, 2-butanone, 4-4-methoxyphenyl, anisylacetone, p, 4-p-methoxyphenyl-2-butanone, methyl oxanone, 1-4-methoxyphenyl-3-butanone
61007
4-(4-methoxyphenyl)butan-2-one
COC1=CC=C(CCC(C)=O)C=C1

4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 7°C to 9°C
Density 1.046
Boiling Point 147°C to 148°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.52
Quantity 10 g
Beilstein 1869592
Solubility Information Not miscible in water.
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material 4- (4-Methoxyphenyl)-2-butanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'