missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Iodophenoxy)tetrahydropyran, 97%, Thermo Scientific™

Catalog Number p-3522506
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
144808-72-0
C11H13IO2
304.127
MFCD09064966
PQUFKWFIWPZFBM-UHFFFAOYSA-N
4-4-iodophenoxy tetrahydropyran, 4-4-iodophenoxy oxane, 4-4-iodophenoxy tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-4-iodophenoxy, acmc-20e5io
24229524
4-(4-iodophenoxy)oxane
C1COCCC1OC2=CC=C(C=C2)I
This item is not returnable. View return policy
144808-72-0
C11H13IO2
304.127
MFCD09064966
PQUFKWFIWPZFBM-UHFFFAOYSA-N
4-4-iodophenoxy tetrahydropyran, 4-4-iodophenoxy oxane, 4-4-iodophenoxy tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-4-iodophenoxy, acmc-20e5io
24229524
4-(4-iodophenoxy)oxane
C1COCCC1OC2=CC=C(C=C2)I
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 304.12
Percent Purity 97%
Chemical Name or Material 4-(4-Iodophenoxy)tetrahydropyran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'