missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Iodobenzyl)morpholine, ≥97%, Thermo Scientific™

Catalog Number p-3075549
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
299159-27-6
C11H14INO
303.143
MFCD04974051
IYDFKIKPNVVXQU-UHFFFAOYSA-N
4-4-iodobenzyl morpholine, 4-4-iodophenyl methyl morpholine, 4-4-iodo-benzyl-morpholine, 4-iodobenzyl morpholine, 4-morpholinomethyliodobenzene
2795500
4-[(4-iodophenyl)methyl]morpholine
C1COCCN1CC2=CC=C(C=C2)I
This item is not returnable. View return policy
299159-27-6
C11H14INO
303.143
MFCD04974051
IYDFKIKPNVVXQU-UHFFFAOYSA-N
4-4-iodobenzyl morpholine, 4-4-iodophenyl methyl morpholine, 4-4-iodo-benzyl-morpholine, 4-iodobenzyl morpholine, 4-morpholinomethyliodobenzene
2795500
4-[(4-iodophenyl)methyl]morpholine
C1COCCN1CC2=CC=C(C=C2)I
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 303.14
Percent Purity 95%
Chemical Name or Material 4-(4-Iodobenzyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'