missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-4-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7055340
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3888-65-1
C11H14FNO
195.24
MFCD00023146
QXWRXWPNHLIZBV-UHFFFAOYSA-N
77497
4-(4-fluorophenyl)piperidin-4-ol
OC1(CCNCC1)C1=CC=C(F)C=C1
This item is not returnable. View return policy
3888-65-1
C11H14FNO
195.24
MFCD00023146
QXWRXWPNHLIZBV-UHFFFAOYSA-N
77497
4-(4-fluorophenyl)piperidin-4-ol
OC1(CCNCC1)C1=CC=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Synonym 4-4-fluorophenyl piperidin-4-ol, 4-piperidinol, 4-4-fluorophenyl, 4-4-fluorophenyl-4-hydroxypiperidine, 4-4-fluorophenyl-4-piperidinol, 4-fluorophenyl-4-hydroxylpiperidinehydrochloride, 4-4-fluoro-phenyl piperidin-4-ol, 4-piperidinol,4-4-fluorophenyl, 4-4-fluorophenyl-4hydroxypiperidine, 4-hydroxy-4-4-fluorophenyl piperidine, 4-4-fluorophenyl-4-hydroxy piperidine
Formula Weight 195.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Fluorophenyl)-4-hydroxypiperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'