missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-2-pyrrolidinone, 95%, Thermo Scientific Chemicals
62.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications
Melting Point | 38°C to 40°C |
Color | White to Cream |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Synonym | 4,4-dimethyl-2-pyrrolidinone, 2-pyrrolidinone, 4,4-dimethyl, 4,4-dimethylpyrrolidine-2-one, 2-pyrrolidone,4,4-dimethyl, 4,4-dimethyl-2-pyrrolidone, 4,4-dimethyl-pyrrolidin-2-one, 4, 4-dimethyl-2-pyrrolidinone |
Solubility Information | Sparingly soluble in water.(0.26g/L) (25°C), |
Formula Weight | 113.2 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 4,4-Dimethyl-2-pyrrolidinone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'