missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxystilbene, 99%, Thermo Scientific Chemicals

Catalog Number 10407880
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 4705-34-4
CAS Min % 98.5
CAS Max % 100.0
Melting Point 213°C to 215°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C16H16O2
Linear Formula CH3OC6H4CH=CHC6H4OCH3
MDL Number MFCD00008414
Quantity 10 g
Beilstein 06, 1023
Synonym bianisal, 4,4'-dimethoxystilbene, bianisylidene, photoanethole, p,p'-dimethoxystilbene, 4,4'-dimethoxy-trans-stilbene, e-1,2-bis 4-methoxyphenyl ethene, 1-methoxy-4-e-2-4-methoxyphenyl ethenyl benzene, 4,4/'-dimethoxystilbene, 1,1'-e-ethene-1,2-diylbis 4-methoxybenzene
InChI Key CAWFCZIEFIQKRV-ONEGZZNKSA-N
SMILES COC1=CC=C(C=C1)C=CC2=CC=C(C=C2)OC
IUPAC Name 1-methoxy-4-[(E)-2-(4-methoxyphenyl)ethenyl]benzene
Molecular Weight (g/mol) 240.3
PubChem CID 641296
Formula Weight 240.3
Percent Purity 99%
Physical Form Glistening Flakes
Chemical Name or Material 4, 4'-Dimethoxystilbene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Dimethoxystilbene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'