missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzophenone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081470
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
90-96-0
C15H14O3
242.27
MFCD00008404
RFVHVYKVRGKLNK-UHFFFAOYSA-N
4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
7032
bis(4-methoxyphenyl)methanone
COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1
This item is not returnable. View return policy
90-96-0
C15H14O3
242.27
MFCD00008404
RFVHVYKVRGKLNK-UHFFFAOYSA-N
4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
7032
bis(4-methoxyphenyl)methanone
COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1

4,4'-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is insoluble in water, soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 142°C to 146°C
Quantity 25 g
Beilstein 1878026
Solubility Information It is insoluble in water,soluble in ethanol.
Formula Weight 242.27
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dimethoxybenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'