missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzophenone, 97%, Thermo Scientific Chemicals

Catalog Number p-114725
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
90-96-0
C15H14O3
242.27
MFCD00008404
RFVHVYKVRGKLNK-UHFFFAOYSA-N
4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
7032
bis(4-methoxyphenyl)methanone
COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1
This item is not returnable. View return policy
90-96-0
C15H14O3
242.27
MFCD00008404
RFVHVYKVRGKLNK-UHFFFAOYSA-N
4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
7032
bis(4-methoxyphenyl)methanone
COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 141.0°C to 146.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula (CH3OC6H4)2CO
Quantity 25 g
Beilstein 08, 317
Formula Weight 242.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4,4′-Dimethoxybenzophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Dimethoxybenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'