missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dihydroxydiphenylmethane, 98%, Thermo Scientific Chemicals
14.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
4,4'-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.
Solubility
It is soluble in ethanol, ether, chloroform, alkali; slightly soluble in DMSO, insoluble in carbon disulfide and water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions.
4,4′-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.
Solubility
It is soluble in ethanol, ether, chloroform, alkali; slightly soluble in DMSO, insoluble in carbon disulfide and water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | 159°C to 163°C |
Quantity | 1 g |
Beilstein | 2049425 |
Solubility Information | It is soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO,insoluble in carbon disulfide and water. |
Formula Weight | 200.24 |
Percent Purity | 98% |
Chemical Name or Material | 4,4'-Dihydroxydiphenylmethane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'