missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl, 99%, ACROS Organics™

Brand:  Acros Organics 212085000

Code : 29

Additional Details : CAS Number : 92-86-4 Weight : 0.58000kg Transport : UN number : 3152 Chem class : 9 Pack group : II


Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10551461

  • £114.60 / 500g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

4, 4'-Dibromobiphenyl
92-86-4
100.0
99%
BrC6H4C6H4Br
MFCD00000101
99%
98.5
≥98.5%
C12H8Br2
312

Specifications

4, 4'-Dibromobiphenyl
165° to 169°C
99%
Authentic
100.0
Crystalline Powder or Crystals
99%
BrC6H4C6H4Br
MFCD00000101
White to light yellow-beige
500g
92-86-4
98.5
Plastic bottle
≥98.5%
C12H8Br2
312
05, 580
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4,4'-Dibromobiphenyl

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS