Alfa Aesar™ 4,4'-Diacetylbiphenyl dioxime, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65777.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15432629

Quantity Price
1 £ 12.4 / 250mg
Estimated Shipment date
from Supplier 07-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4,4'-Diacetylbiphenyl dioxime
Formula Weight 268.32
MDL Number MFCD25977523

Specification

Formula Weight 268.32
Assay 97%
Quantity 250mg
Color White to pale brown
Chemical Name or Material 4,4'-Diacetylbiphenyl dioxime
MDL Number MFCD25977523
Physical Form Solid
Documentation

Technical documents