missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98%, ACROS Organics™

Brand:  Acros Organics 305440050

Code : 29

Additional Details : CAS Number : 787-69-9 Weight : 0.05500kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11984851

  • £62.00 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

4, 4'-Diacetylbiphenyl
97.5
98%
C16H14O2
238.28
787-69-9
100.0
98%
CH3COC6H4C6H4COCH3

Specifications

4, 4'-Diacetylbiphenyl
787-69-9
100.0
98%
C16H14O2
238.28
Glass bottle
192.0° to 196.0°C
5g
97.5
Crystalline Powder
98%
CH3COC6H4C6H4COCH3
Authentic
Colorless to light tan
Videos
SDS