missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-tert-butylbiphenyl, 99+%, Thermo Scientific Chemicals

Catalog Number p-3757475
86.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1625-91-8
C20H26
266.42
MFCD00008834
CDKCEZNPAYWORX-UHFFFAOYSA-N
4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
74195
1-tert-butyl-4-(4-tert-butylphenyl)benzene
CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1625-91-8
C20H26
266.42
MFCD00008834
CDKCEZNPAYWORX-UHFFFAOYSA-N
4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
74195
1-tert-butyl-4-(4-tert-butylphenyl)benzene
CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 126°C to 129°C
Color Orange-Yellow to Yellow
Boiling Point 190°C to 192°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4C6H4C(CH3)3
Quantity 25 g
Beilstein 05, 298
Solubility Information Solubility in water: .. Other solubilities: soluble in dioxane
Formula Weight 266.42
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Di-tert-butylbiphenyl
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'