missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-chlorophenyl)piperidin-4-ol, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 100 g |
Formula Weight | 211.69 |
Percent Purity | 97% |
Chemical Name or Material | 4-(4-Chlorophenyl)piperidin-4-ol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'