missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(4-Chlorobenzyl)pyridine, 97%

Brand:  Alfa Aesar™ A10673.36

Code : NEW

Additional Details : CAS Number : 4409-11-4 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 11403434

  • £98.80 / 500g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

4- (4-Chlorobenzyl)pyridine
C12H10ClN
MFCD00006444
4409-11-4
203.67
1367705

Specifications

4- (4-Chlorobenzyl)pyridine
123° to 125°C (6mmHg)
4409-11-4
C12H10ClN
MFCD00006444
>110°C (230°F)
500g
97%
203.67
1367705
Videos
SDS