missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)-1,2,3-thiadiazole, 97%, Maybridge

Brand:  Maybridge CC16410EA

Code : 31

Additional Details : CAS Number : 40753-13-7 Weight : 0.06000kg

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10522842

  • £104.00 / 10g


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(4-Bromophenyl)-1, 2, 3-thiadiazole
96.5
97%
C8H5BrN2S
MFCD00084907
40753-13-7
100.0
97%
241.11

Specifications

4-(4-Bromophenyl)-1, 2, 3-thiadiazole
96.5
Amber Glass Bottle
97%
C8H5BrN2S
MFCD00084907
40753-13-7
100.0
10g
97%
241.11
Videos
SDS