missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Azodianiline, 95%, Thermo Scientific Chemicals

Catalog Number p-61510
128.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
538-41-0
C12H12N4
212.26
MFCD00041892
KQIKKETXZQDHGE-UHFFFAOYSA-N
4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
10855
4-[(4-aminophenyl)diazenyl]aniline
NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
538-41-0
C12H12N4
212.26
MFCD00041892
KQIKKETXZQDHGE-UHFFFAOYSA-N
4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
10855
4-[(4-aminophenyl)diazenyl]aniline
NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 245°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 2979
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,very soluble in benzene and chloroform
Formula Weight 212.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4, 4'-Azodianiline, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Azodianiline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'