missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane, ≥97%, Maybridge™
£81.37 - £203.00
Chemical Identifiers
CAS | 912569-68-7 |
---|---|
Molecular Formula | C19H23BO3 |
Molecular Weight (g/mol) | 310.2 |
MDL Number | MFCD09065022 |
InChI Key | AFDZGEOQKHIWKF-UHFFFAOYSA-N |
Synonym | 4,4,5,5-tetramethyl-2-3-phenoxymethyl phenyl-1,3,2-dioxaborolane |
PubChem CID | 24229702 |
IUPAC Name | 4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)COC3=CC=CC=C3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10728835
|
Maybridge
CC63639CB |
250mg | Amber Glass Bottle |
£81.37
250mg |
Estimated Shipment 26-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! |
10002214
|
Maybridge
CC63639DA |
1g | Amber Glass Bottle |
£203.00
1g |
Estimated Shipment 26-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! |
Chemical Identifiers
912569-68-7 | |
310.2 | |
AFDZGEOQKHIWKF-UHFFFAOYSA-N | |
24229702 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)COC3=CC=CC=C3 |
C19H23BO3 | |
MFCD09065022 | |
4,4,5,5-tetramethyl-2-3-phenoxymethyl phenyl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane |
Specifications
4,4,5,5-Tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane | |
250mg | |
96.5 | |
97+% | |
MFCD09065022 | |
AFDZGEOQKHIWKF-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane | |
24229702 | |
97% |
Amber Glass Bottle | |
912569-68-7 | |
100.0 | |
C19H23BO3 | |
4,4,5,5-tetramethyl-2-3-phenoxymethyl phenyl-1,3,2-dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)COC3=CC=CC=C3 | |
310.2 | |
310.2 |