missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5,5-Pentafluoropentan-1-ol, 95%, Thermo Scientific Chemicals

Catalog Number 11994611
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
148043-73-6
C5H7F5O
178.1
QROUUECTKRZFHF-UHFFFAOYSA-N
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
547967
4,4,5,5,5-pentafluoropentan-1-ol
C(CC(C(F)(F)F)(F)F)CO
This item is not returnable. View return policy
148043-73-6
C5H7F5O
178.1
QROUUECTKRZFHF-UHFFFAOYSA-N
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
547967
4,4,5,5,5-pentafluoropentan-1-ol
C(CC(C(F)(F)F)(F)F)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Boiling Point 134°C to 135°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 178.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4,4,5,5,5-Pentafluoropentan-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 4,4,5,5,5-Pentafluoropentan-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'